Gene Transcripts
- Gene Symbol
- Rbm25
- Entrez Gene ID
- 67039
- Gene Name
- RNA binding motif protein 25
- Gene Aliases
- 2600011C06Rik, 2610015J01Rik, A130095G20Rik, AI159652, AL023075, AU043498, RNPC7
- Chromosome Location
- Chr. 12: 83632204 - 83683127 on Build GRCm38
- UniGene ID
- Mm.46005
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK038317.1 |
- |
1 |
349 |
|
BC010792.1 |
- |
8 |
2096 |
|
BC023749.1 |
- |
7 |
2059 |
|
View Full Product Details