Gene Transcripts

Gene Symbol
Fmo5
Entrez Gene ID
246248
Gene Name
flavin containing monooxygenase 5
Gene Aliases
Chromosome Location
Chr. 2: 199796870 - 199823927 on Build Rnor_6.0
UniGene ID
Rn.7038
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144739.1 NP_653340.1 6 1137
GenBank mRNA AF458413.1 - 6 1137
BC070883.1 - 6 865 7096939

View Full Product Details