Gene Transcripts

Gene Symbol
KMT2C
Entrez Gene ID
58508
Gene Name
lysine methyltransferase 2C
Gene Aliases
HALR, MLL3
Chromosome Location
Chr. 7: 152134925 - 152436642 on Build GRCh38
UniGene ID
Hs.647120
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_170606.2 NP_733751.2 34 5255
XM_005250025.4 XP_005250082.1 36 5109
XM_005250026.3 XP_005250083.1 36 5143
XM_005250027.4 XP_005250084.1 35 5317
XM_005250028.4 XP_005250085.1 35 5317
XM_005250031.4 XP_005250088.1 35 5317
XM_006716077.3 XP_006716140.1 35 5317
XM_006716078.3 XP_006716141.1 35 5317
XM_006716079.3 XP_006716142.1 35 5317
XM_011516450.2 XP_011514752.1 34 5269
XM_011516451.2 XP_011514753.1 34 5197
XM_011516452.2 XP_011514754.1 33 5164
XM_011516453.2 XP_011514755.1 35 5317
XM_011516454.2 XP_011514756.1 30 4312
XM_011516456.2 XP_011514758.1 34 5269
XM_017012480.1 XP_016867969.1 36 5146
XM_017012481.1 XP_016867970.1 36 5106
XM_017012482.1 XP_016867971.1 35 5317
XM_017012483.1 XP_016867972.1 35 5317
XM_017012484.1 XP_016867973.1 35 5284
XM_017012485.1 XP_016867974.1 34 5266
XM_017012486.1 XP_016867975.1 35 5317
XM_017012487.1 XP_016867976.1 35 5285
XM_017012488.1 XP_016867977.1 35 5317
XM_017012489.1 XP_016867978.1 16 1824
XM_017012490.1 XP_016867979.1 13 1396
GenBank mRNA AB040939.2 - 23 3371
AF264750.2 - 20 2582
AY024361.1 - 34 5216
BC151837.1 - 23 3371

View Full Product Details