Gene Transcripts

Gene Symbol
CD200R1
Entrez Gene ID
131450
Gene Name
CD200 receptor 1
Gene Aliases
CD200R, HCRTR2, MOX2R, OX2R
Chromosome Location
Chr. 3: 112922685 - 112975090 on Build GRCh38
UniGene ID
Hs.309158
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138806.3 NP_620161.1 8 2209
NM_170780.2 NP_740750.1 7 2140
GenBank mRNA AK126349.1 - 7 2140
AY358910.1 - 8 2234

View Full Product Details