Gene Transcripts
- Gene Symbol
- DNMT3A
- Entrez Gene ID
- 1788
- Gene Name
- DNA methyltransferase 3 alpha
- Gene Aliases
- DNMT3A2, M.HsaIIIA, TBRS
- Chromosome Location
- Chr. 2: 25232961 - 25342590 on Build GRCh38
- UniGene ID
- Hs.515840
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001320893.1 |
NP_001307822.1
|
14, 15 |
1903 |
|
NM_022552.4 |
NP_072046.2
|
19, 20 |
2589 |
|
NM_153759.3 |
NP_715640.2
|
15, 16 |
1873 |
|
NM_175629.2 |
NP_783328.1
|
19, 20 |
2660 |
|
XM_005264175.4 |
XP_005264232.1
|
19, 20 |
2523 |
|
XM_005264177.4 |
XP_005264234.1
|
14, 15 |
1815 |
|
XM_011532662.2 |
XP_011530964.1
|
17, 18 |
2525 |
|
XM_011532663.2 |
XP_011530965.1
|
20, 21 |
2702 |
|
XM_011532666.2 |
XP_011530968.1
|
15, 16 |
1888 |
|
XM_011532667.2 |
XP_011530969.1
|
14, 15 |
1705 |
|
XM_017003526.1 |
XP_016859015.1
|
19, 20 |
2631 |
|
XM_017003527.1 |
XP_016859016.1
|
14, 15 |
1859 |
|
GenBank mRNA |
AF067972.2 |
- |
19, 20 |
2551 |
|
AF331856.1 |
- |
19, 20 |
2539 |
|
AF480163.1 |
- |
15, 16 |
1869 |
|
AK025230.1 |
- |
9, 10 |
913 |
|
BC018214.1 |
- |
4, 5 |
442 |
|
BC023612.2 |
- |
5, 6 |
641 |
|
BC043617.1 |
- |
19, 20 |
2559 |
6150112 |
W76111.1 |
- |
4, 5 |
411 |
|
View Full Product Details