Gene Transcripts

Gene Symbol
ZFP64
Entrez Gene ID
55734
Gene Name
ZFP64 zinc finger protein
Gene Aliases
ZNF338
Chromosome Location
Chr. 20: 52084011 - 52191985 on Build GRCh38
UniGene ID
Hs.473082
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319146.1 NP_001306075.1 5 869
NM_018197.2 NP_060667.2 6 1163
NM_022088.4 NP_071371.3 5 1001
NM_199426.1 NP_955458.1 6 1157
NM_199427.2 NP_955459.2 9 1628
XM_005260449.1 XP_005260506.1 5 580
XM_006723822.1 XP_006723885.1 5 798
XM_011528900.1 XP_011527202.1 7 1054
XM_017027945.1 XP_016883434.1 9 1414
XM_017027946.1 XP_016883435.1 5 723
XM_017027947.1 XP_016883436.1 8 1270
XM_017027948.1 XP_016883437.1 6 702
XM_017027949.1 XP_016883438.1 7 1021
GenBank mRNA AK001596.1 - 6 953
AK001744.1 - 5 728
AK022690.1 - 5 869
AK090547.1 - 7 825
BC012759.2 - 6 1163 3634346
BC021087.1 - 9 1349
BC041622.1 - 6 1163 3633938
BT009760.1 - 9 1279

View Full Product Details