Gene Transcripts
- Gene Symbol
- XCL1
- Entrez Gene ID
- 6375
- Gene Name
- X-C motif chemokine ligand 1
- Gene Aliases
- ATAC, LPTN, LTN, SCM-1, SCM-1a, SCM1, SCM1A, SCYC1
- Chromosome Location
- Chr. 1: 168574128 - 168582077 on Build GRCh38
- UniGene ID
- Hs.546295
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
U23772.1 |
- |
3 |
403 |
|
- Gene Symbol
- XCL2
- Entrez Gene ID
- 6846
- Gene Name
- X-C motif chemokine ligand 2
- Gene Aliases
- SCM-1b, SCM1B, SCYC2
- Chromosome Location
- Chr. 1: 168540765 - 168543997 on Build GRCh38
- UniGene ID
- Hs.458346
- Species
- Homo sapiens
View Full Product Details