Gene Transcripts

Gene Symbol
TGIF2LX
Entrez Gene ID
90316
Gene Name
TGFB induced factor homeobox 2 like, X-linked
Gene Aliases
TGIFLX
Chromosome Location
Chr. X: 89921941 - 89922883 on Build GRCh38
UniGene ID
Hs.592220
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138960.3 NP_620410.3 2 621
GenBank mRNA AF497480.1 - 1 601
BC101144.2 - 1 606
BC101145.1 - 1 606
BC117398.1 - 2 621 40125949
BC117400.1 - 2 621 40125951

View Full Product Details