Gene Transcripts
- Gene Symbol
- TDGF1
- Entrez Gene ID
- 6997
- Gene Name
- teratocarcinoma-derived growth factor 1
- Gene Aliases
- CR, CRGF, CRIPTO
- Chromosome Location
- Chr. 3: 46574555 - 46582463 on Build GRCh38
- UniGene ID
- Hs.385870
- Species
- Homo sapiens
- Gene Symbol
- TDGF1P3
- Entrez Gene ID
- 6998
- Gene Name
- teratocarcinoma-derived growth factor 1 pseudogene 3
- Gene Aliases
- CR-3, CRIPTO, CRIPTO-3, CRIPTO3, TDGF1, TDGF2, TDGF3
- Chromosome Location
- Chr. X: 110520312 - 110523021 on Build GRCh38
- UniGene ID
- Hs.592361
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002718.2 |
|
1 |
1337 |
|
GenBank mRNA |
M96956.1 |
- |
1 |
1328 |
|
View Full Product Details