Gene Transcripts

Gene Symbol
CCL4L2
Entrez Gene ID
9560
Gene Name
C-C motif chemokine ligand 4 like 2
Gene Aliases
AT744.2, CCL4L, SCYA4L, SCYQ4L2
Chromosome Location
Chr. 17: 36211063 - 36212878 on Build GRCh38
UniGene ID
Hs.515183
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291468.1 NP_001278397.1 3 642
NM_001291469.1 NP_001278398.1 3 846
NM_001291470.1 NP_001278399.1 2 1248
NM_001291471.1 NP_001278400.1 3 826
NM_001291472.1 NP_001278401.1 3 805
NM_001291473.1 NP_001278402.1 3 775
NM_001291474.1 NP_001278403.1 2 731
NM_001291475.1 NP_001278404.1 3 836
NR_111970.1 2 527
GenBank mRNA AY766450.1 - 3 642
AY766451.1 - 3 846
AY766452.1 - 2 1248
AY766453.1 - 3 826
AY766454.1 - 3 805
AY766455.1 - 3 775
AY766456.1 - 2 527
AY766457.1 - 2 731
AY766458.1 - 3 836

Gene Symbol
CCL4L1
Entrez Gene ID
388372
Gene Name
C-C motif chemokine ligand 4 like 1
Gene Aliases
AT744.2, CCL4L, LAG-1, LAG1, MIP-1-beta, SCYA4L, SCYA4L1, SCYA4L2
Chromosome Location
Chr. : - on Build
UniGene ID
Hs.449862
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207007.3 NP_996890.1 3 812
NR_111969.1 2 697
GenBank mRNA AY766447.1 - 3 657
AY766449.1 - 2 542

View Full Product Details