Gene Transcripts

Gene Symbol
LOXHD1
Entrez Gene ID
125336
Gene Name
lipoxygenase homology domains 1
Gene Aliases
DFNB77, LH2D1
Chromosome Location
Chr. 18: 46476961 - 46657115 on Build GRCh38
UniGene ID
Hs.345877
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145472.2 NP_001138944.1 16 2405
NM_001145473.2 NP_001138945.1 2 266
NM_001173129.1 NP_001166600.1 2 266
NM_001308013.1 NP_001294942.1 14 2059
NM_144612.6 NP_653213.6 33 5143
XM_006722388.3 XP_006722451.1 15 2239
XM_006722389.3 XP_006722452.1 16 2415
XM_006722390.3 XP_006722453.1 16 2483
XM_006722391.3 XP_006722454.1 14 2053
XM_011525804.2 XP_011524106.1 22 3563
XM_011525807.2 XP_011524109.1 13 1949
XM_011525810.2 XP_011524112.1 2 486
XM_011525811.2 XP_011524113.1 2 1051
XM_017025540.1 XP_016881029.1 26 4359
XM_017025541.1 XP_016881030.1 27 4446
XM_017025542.1 XP_016881031.1 27 4446
XM_017025543.1 XP_016881032.1 27 4446
XM_017025544.1 XP_016881033.1 27 4446
XM_017025545.1 XP_016881034.1 27 4446
XM_017025548.1 XP_016881037.1 32 4793
GenBank mRNA AK057232.1 - 2 253
AK127869.1 - 26 4456
AK302484.1 - 14 2050
BC041860.1 - 16 2387 5271046
BC047720.1 - 2 266
BX647229.1 - 14 2059

View Full Product Details