Gene Transcripts

Gene Symbol
Cryzl1
Entrez Gene ID
288256
Gene Name
crystallin, zeta (quinone reductase)-like 1
Gene Aliases
Chromosome Location
Chr. 11: 31847741 - 31893147 on Build Rnor_6.0
UniGene ID
Rn.145951
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013044.1 NP_001013062.1 13 1430
GenBank mRNA BC083853.1 - 13 1430

View Full Product Details