Gene Transcripts
- Gene Symbol
- Vmn1r121
- Entrez Gene ID
- 667240
- Gene Name
- vomeronasal 1 receptor 121
- Gene Aliases
- EG667240, Gm8533
- Chromosome Location
- Chr. 7: 21097569 - 21098513 on Build GRCm38
- UniGene ID
- Mm.480259
- Species
- Mus musculus
- Gene Symbol
- Gm10665
- Entrez Gene ID
- 100043058
- Gene Name
- predicted gene 10665
- Gene Aliases
- ENSMUSG00000074308, Vmn1r150
- Chromosome Location
- Chr. 7: 22463110 - 22464075 on Build GRCm38
- UniGene ID
- Mm.484986
- Species
- Mus musculus
- Gene Symbol
- Gm10670
- Entrez Gene ID
- 100042971
- Gene Name
- predicted gene 10670
- Gene Aliases
- ENSMUSG00000074330, Vmn1r102
- Chromosome Location
- Chr. 7: 20473960 - 20474925 on Build GRCm38
- UniGene ID
- Mm.389840
- Species
- Mus musculus
- Gene Symbol
- Gm4207
- Entrez Gene ID
- 100043069
- Gene Name
- vomeronasal 1 receptor Vmn1r-ps81 pseudogene
- Gene Aliases
- Vmn1r-ps81
- Chromosome Location
- Chr. 7: 22806346 - 22808474 on Build GRCm38
- UniGene ID
- Mm.431911
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC107205.1 |
- |
1 |
357 |
|
BC119491.1 |
- |
|
357 |
|
- Gene Symbol
- Vmn1r-ps79
- Entrez Gene ID
- 100042966
- Gene Name
- vomeronasal 1 receptor, pseudogene 79
- Gene Aliases
- Gm652
- Chromosome Location
- Chr. 7: 20356626 - 20358918 on Build GRCm38
- UniGene ID
- Mm.431911
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC115870.1 |
- |
1 |
1417 |
|
BC119492.1 |
- |
1 |
357 |
|
View Full Product Details