Gene Transcripts

Gene Symbol
Thumpd1
Entrez Gene ID
309041
Gene Name
THUMP domain containing 1
Gene Aliases
Chromosome Location
Chr. 1: 189457582 - 190370515 on Build Rnor_6.0
UniGene ID
Rn.12345
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009688.1 NP_001009688.1 4 2356
GenBank mRNA BC087659.1 - 4 2356

View Full Product Details