Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|---|
RefSeq | NM_001035235.3 | NP_001030312.2 | 3 | 848 | ||
NR_045586.1 | 3 | 908 | ||||
NR_045587.1 | 2 | 366 | ||||
GenBank mRNA | AF092038.1 | - | 3 | 210 | ||
AF293024.1 | - | LIT1246 SRA |
3 | 432 | ||
AF293025.1 | - | LIT1250 SRA |
3 | 432 | ||
AF293026.1 | - | LIT1251 SRA |
3 | 432 | ||
AF318361.1 | - | 3 | 849 | |||
AK054960.1 | - | 3 | 1065 | |||
BC040043.2 | - | 2 | 180 | |||
CX165918.1 | - | 3 | 470 | |||
DA311407.1 | - | 2 | 344 | |||
DQ286291.1 | - | 3 | 492 |