Gene Transcripts
- Gene Symbol
- BC061212
- Entrez Gene ID
- 381724
- Gene Name
- cDNA sequence BC061212
- Gene Aliases
- Chromosome Location
- Chr. 5: 94066454 - 94070545 on Build GRCm38
- UniGene ID
- Mm.380154
- Species
- Mus musculus
- Gene Symbol
- BC080696
- Entrez Gene ID
- 665521
- Gene Name
- cDNA sequence BC080696
- Gene Aliases
- Chromosome Location
- Chr. 5: 94434131 - 94436940 on Build GRCm38
- UniGene ID
- Mm.380154
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC080696.1 |
- |
3 |
1180 |
|
- Gene Symbol
- Gm7978
- Entrez Gene ID
- 666203
- Gene Name
- predicted gene 7978
- Gene Aliases
- EG666203
- Chromosome Location
- Chr. 5: 95732092 - 95736173 on Build GRCm38
- UniGene ID
- Mm.380154
- Species
- Mus musculus
View Full Product Details