Gene Transcripts

Gene Symbol
ETNPPL
Entrez Gene ID
64850
Gene Name
ethanolamine-phosphate phospho-lyase
Gene Aliases
AGXT2L1
Chromosome Location
Chr. 4: 108742040 - 108763180 on Build GRCh38
UniGene ID
Hs.106576
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146590.1 NP_001140062.1 9 1224
NM_001146627.1 NP_001140099.1 8 1145
NM_031279.3 NP_112569.2 9 1242
NR_027474.1 9 1264
NR_027475.1 8 1123
XM_006714290.2 XP_006714353.1 9 1993
GenBank mRNA AJ298293.1 - 9 1152
AK091888.1 - 9 1061
AK294072.1 - 8 1030
AK294235.1 - 9 1238
BC022526.1 - 9 1216 4797767

View Full Product Details