Gene Transcripts

Gene Symbol
Sdc1
Entrez Gene ID
25216
Gene Name
syndecan 1
Gene Aliases
HSPG, SYNDECA, Synd1, Syndecan
Chromosome Location
Chr. 6: 33885576 - 33908038 on Build Rnor_6.0
UniGene ID
Rn.11176
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013026.2 NP_037158.1 3 414
GenBank mRNA BC061834.1 - 3 414
M81785.1 - 3 429
S61865.1 - 3 429
X60651.1 - 2 104

View Full Product Details