Gene Transcripts
- Gene Symbol
- Htr5a
- Entrez Gene ID
- 25689
- Gene Name
- 5-hydroxytryptamine (serotonin) receptor 5A, G protein-coupled
- Gene Aliases
- 5HT5A, MR22
- Chromosome Location
- Chr. 4: 199916 - 209599 on Build Rnor_6.0
- UniGene ID
- Rn.10569
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013148.1 |
NP_037280.1
|
2 |
979 |
|
GenBank mRNA |
L10072.1 |
- |
2 |
979 |
|
View Full Product Details