Gene Transcripts
- Gene Symbol
- BRAF
- Entrez Gene ID
- 673
- Gene Name
- B-Raf proto-oncogene, serine/threonine kinase
- Gene Aliases
- B-RAF1, B-raf, BRAF1, NS7, RAFB1
- Chromosome Location
- Chr. 7: 140719331 - 140924764 on Build GRCh38
- UniGene ID
- Hs.550061
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004333.4 |
NP_004324.2
|
1, 2 |
207 |
|
XM_005250045.2 |
XP_005250102.1
|
1, 2 |
154 |
|
XM_017012558.1 |
XP_016868047.1
|
1, 2 |
154 |
|
XM_017012559.1 |
XP_016868048.1
|
1, 2 |
154 |
|
GenBank mRNA |
AI203435.1 |
- |
1, 2 |
179 |
|
AK314346.1 |
- |
1, 2 |
154 |
|
BC101757.1 |
- |
1, 2 |
168 |
8069263 |
BC112079.1 |
- |
1, 2 |
168 |
8327547 |
DB454480.1 |
- |
1, 2 |
372 |
|
DN996440.1 |
- |
1, 2 |
405 |
|
M95712.2 |
- |
1, 2 |
207 |
|
View Full Product Details