Gene Transcripts
- Gene Symbol
- Cdk16
- Entrez Gene ID
- 81741
- Gene Name
- cyclin-dependent kinase 16
- Gene Aliases
- Pctk1
- Chromosome Location
- Chr. X: 1707285 - 1718821 on Build Rnor_6.0
- UniGene ID
- Rn.16871
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031077.1 |
NP_112339.1
|
3, 4 |
725 |
|
GenBank mRNA |
U36444.1 |
- |
3, 4 |
725 |
|
View Full Product Details