Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005654.5 | NP_005645.1 | 1 | 2042 | |
GenBank mRNA | BC004154.2 | - | 1 | 443 | |
BC017493.1 | - | 1 | 2042 | 3912075 | |
HQ709180.1 | - | 2 | 280 | ||
HQ709181.1 | - | 2 | 202 | ||
HQ709182.1 | - | 2 | 232 | ||
HQ709183.1 | - | 2 | 280 | ||
X16155.1 | - | 1 | 340 |