Gene Transcripts

Gene Symbol
Capn6
Entrez Gene ID
12338
Gene Name
calpain 6
Gene Aliases
Chromosome Location
Chr. X: 143802236 - 143827412 on Build GRCm38
UniGene ID
Mm.30290
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007603.3 NP_031629.3 2 294
GenBank mRNA AK048028.1 - 2 215
AK145116.1 - 2 309
CN700442.1 - 2 294
Y12582.1 - 2 203

View Full Product Details