Gene Transcripts
- Gene Symbol
- Ncor1
- Entrez Gene ID
- 20185
- Gene Name
- nuclear receptor co-repressor 1
- Gene Aliases
- 5730405M06Rik, A230020K14Rik, N-CoR, RIP13, Rxrip13, mKIAA1047
- Chromosome Location
- Chr. 11: 62315643 - 62457332 on Build GRCm38
- UniGene ID
- Mm.271814
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
U35312.1 |
- |
2 |
378 |
|
View Full Product Details