Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001161420.1 | NP_001154892.1 | 7, 8 | 1662 | |
NM_013703.2 | NP_038731.2 | 7, 8 | 1662 | ||
GenBank mRNA | AK076172.1 | - | 7, 8 | 1667 | |
AK136838.1 | - | 7, 8 | 1482 | ||
BC013622.1 | - | 7, 8 | 1098 | 3968213 | |
L33417.1 | - | 7, 8 | 1086 | ||
U06670.1 | - | 7, 8 | 1248 |