Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|---|
RefSeq | NM_025291.3 | NP_079567.2 | 4 | 401 | ||
GenBank mRNA | AF092039.1 | - | LIT1243 SRA |
3 | 305 | |
AK007302.1 | - | 4 | 387 | |||
AK010375.1 | - | 4 | 387 | |||
AK154232.1 | - | 4 | 371 | |||
BC026480.1 | - | 2 | 180 | |||
BC048362.1 | - | 4 | 373 | 2646489 | ||
BQ748094.1 | - | 2 | 69 | |||
BX516162.1 | - | 3 | 468 |