Gene Transcripts
- Gene Symbol
- Fam192a
- Entrez Gene ID
- 102122
- Gene Name
- family with sequence similarity 192, member A
- Gene Aliases
- 1700001O11Rik, 2310065K24Rik, AI596259, Nip30
- Chromosome Location
- Chr. 8: 94574940 - 94601726 on Build GRCm38
- UniGene ID
- Mm.250425
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_028221.4 |
NP_082497.2
|
4, 5 |
556 |
|
GenBank mRNA |
AK010053.1 |
- |
4, 5 |
521 |
|
AK033988.1 |
- |
4, 5 |
487 |
|
AK050704.1 |
- |
4, 5 |
491 |
|
AK151603.1 |
- |
4, 5 |
510 |
|
BC016088.1 |
- |
3, 4 |
296 |
4503452 |
BC023158.1 |
- |
4, 5 |
395 |
4946201 |
BC025472.1 |
- |
4, 5 |
532 |
5353148 |
BC027060.1 |
- |
4, 5 |
518 |
5358022 |
BC027818.1 |
- |
3, 4 |
324 |
5320967 |
BC040818.1 |
- |
|
509 |
4194531 |
View Full Product Details