Gene Transcripts
- Gene Symbol
- Vmn1r100
- Entrez Gene ID
- 100043536
- Gene Name
- vomeronasal 1 receptor 100
- Gene Aliases
- ENSMUSG00000074331, Gm10671
- Chromosome Location
- Chr. 7: 20417942 - 20418865 on Build GRCm38
- UniGene ID
- Mm.484979
- Species
- Mus musculus
- Gene Symbol
- Gm10666
- Entrez Gene ID
- 100043037
- Gene Name
- predicted gene 10666
- Gene Aliases
- ENSMUSG00000074310, Vmn1r141
- Chromosome Location
- Chr. 7: 22133776 - 22134741 on Build GRCm38
- UniGene ID
- Mm.484998
- Species
- Mus musculus
- Gene Symbol
- Gm16442
- Entrez Gene ID
- 620401
- Gene Name
- predicted gene 16442
- Gene Aliases
- EG667071, Vmn1r92
- Chromosome Location
- Chr. 7: 20121656 - 20122579 on Build GRCm38
- UniGene ID
- Mm.484999
- Species
- Mus musculus
- Gene Symbol
- Gm16451
- Entrez Gene ID
- 100043033
- Gene Name
- predicted pseudogene 16451
- Gene Aliases
- EG667454, Vmn1r140
- Chromosome Location
- Chr. 7: 22117322 - 22118245 on Build GRCm38
- UniGene ID
- Mm.482060
- Species
- Mus musculus
- Gene Symbol
- Gm4133
- Entrez Gene ID
- 100042964
- Gene Name
- predicted gene 4133
- Gene Aliases
- Vmn1r98
- Chromosome Location
- Chr. 7: 20333142 - 20334065 on Build GRCm38
- UniGene ID
- Mm.484995
- Species
- Mus musculus
- Gene Symbol
- Gm4175
- Entrez Gene ID
- 100043019
- Gene Name
- predicted gene 4175
- Gene Aliases
- Vmn1r133
- Chromosome Location
- Chr. 7: 21760531 - 21761475 on Build GRCm38
- UniGene ID
- Mm.484994
- Species
- Mus musculus
- Gene Symbol
- Gm4187
- Entrez Gene ID
- 100043043
- Gene Name
- predicted gene 4187
- Gene Aliases
- Vmn1r146
- Chromosome Location
- Chr. 7: 22328794 - 22329717 on Build GRCm38
- UniGene ID
- Mm.482053
- Species
- Mus musculus
- Gene Symbol
- Gm4207
- Entrez Gene ID
- 100043069
- Gene Name
- vomeronasal 1 receptor Vmn1r-ps81 pseudogene
- Gene Aliases
- Vmn1r-ps81
- Chromosome Location
- Chr. 7: 22806346 - 22808474 on Build GRCm38
- UniGene ID
- Mm.431911
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC107205.1 |
- |
1 |
163 |
|
BC119491.1 |
- |
|
163 |
|
- Gene Symbol
- Gm4214
- Entrez Gene ID
- 100043079
- Gene Name
- predicted gene 4214
- Gene Aliases
- Vmn1r161
- Chromosome Location
- Chr. 7: 22983729 - 22984673 on Build GRCm38
- UniGene ID
- Mm.482051
- Species
- Mus musculus
- Gene Symbol
- Gm4498
- Entrez Gene ID
- 100043523
- Gene Name
- predicted gene 4498
- Gene Aliases
- Vmn1r97
- Chromosome Location
- Chr. 7: 20232908 - 20233828 on Build GRCm38
- UniGene ID
- Mm.482055
- Species
- Mus musculus
- Gene Symbol
- Gm5725
- Entrez Gene ID
- 435940
- Gene Name
- predicted gene 5725
- Gene Aliases
- EG435940, Vmn1r136
- Chromosome Location
- Chr. 7: 21843344 - 21844267 on Build GRCm38
- UniGene ID
- Mm.482057
- Species
- Mus musculus
- Gene Symbol
- Gm5726
- Entrez Gene ID
- 435946
- Gene Name
- predicted gene 5726
- Gene Aliases
- EG435946, Vmn1r105
- Chromosome Location
- Chr. 7: 20544834 - 20545757 on Build GRCm38
- UniGene ID
- Mm.484990
- Species
- Mus musculus
- Gene Symbol
- Gm5728
- Entrez Gene ID
- 435949
- Gene Name
- predicted gene 5728
- Gene Aliases
- EG435949, Vmn1r99
- Chromosome Location
- Chr. 7: 20374493 - 20375410 on Build GRCm38
- UniGene ID
- Mm.485000
- Species
- Mus musculus
- Gene Symbol
- Gm6164
- Entrez Gene ID
- 620574
- Gene Name
- predicted gene 6164
- Gene Aliases
- EG620574, Vmn1r96
- Chromosome Location
- Chr. 7: 20204684 - 20205607 on Build GRCm38
- UniGene ID
- Mm.482065
- Species
- Mus musculus
- Gene Symbol
- Gm8453
- Entrez Gene ID
- 667094
- Gene Name
- predicted gene 8453
- Gene Aliases
- EG667094, Vmn1r145
- Chromosome Location
- Chr. 7: 22228564 - 22229484 on Build GRCm38
- UniGene ID
- Mm.484989
- Species
- Mus musculus
- Gene Symbol
- Gm8653
- Entrez Gene ID
- 667472
- Gene Name
- predicted gene 8653
- Gene Aliases
- EG667472, Vmn1r144
- Chromosome Location
- Chr. 7: 22200341 - 22201264 on Build GRCm38
- Species
- Mus musculus
- Gene Symbol
- Gm8660
- Entrez Gene ID
- 667485
- Gene Name
- predicted gene 8660
- Gene Aliases
- EG667485, Vmn1r147
- Chromosome Location
- Chr. 7: 22370122 - 22371039 on Build GRCm38
- UniGene ID
- Mm.482056
- Species
- Mus musculus
- Gene Symbol
- Gm8677
- Entrez Gene ID
- 667512
- Gene Name
- predicted gene 8677
- Gene Aliases
- EG667512, Vmn1r153
- Chromosome Location
- Chr. 7: 22533964 - 22534887 on Build GRCm38
- UniGene ID
- Mm.482061
- Species
- Mus musculus
- Gene Symbol
- Gm8693
- Entrez Gene ID
- 667543
- Gene Name
- predicted gene 8693
- Gene Aliases
- 100042980, EG667543, Gm4144, Vmn1r108, Vmn1r156
- Chromosome Location
- Chr. 7: 22691397 - 22692317 on Build GRCm38
- UniGene ID
- Mm.482054
- Species
- Mus musculus
- Gene Symbol
- Gm8720
- Entrez Gene ID
- 667599
- Gene Name
- predicted gene 8720
- Gene Aliases
- EG667599, Vmn1r164
- Chromosome Location
- Chr. 7: 23066533 - 23067456 on Build GRCm38
- UniGene ID
- Mm.484993
- Species
- Mus musculus
- Gene Symbol
- Vmn1r-ps79
- Entrez Gene ID
- 100042966
- Gene Name
- vomeronasal 1 receptor, pseudogene 79
- Gene Aliases
- Gm652
- Chromosome Location
- Chr. 7: 20356626 - 20358918 on Build GRCm38
- UniGene ID
- Mm.431911
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC115870.1 |
- |
1 |
1223 |
|
BC119492.1 |
- |
1 |
163 |
|
- Gene Symbol
- Vmn1r101
- Entrez Gene ID
- 100042968
- Gene Name
- vomeronasal 1 receptor 101
- Gene Aliases
- Gm4135
- Chromosome Location
- Chr. 7: 20441670 - 20442593 on Build GRCm38
- UniGene ID
- Mm.480292
- Species
- Mus musculus
- Gene Symbol
- Vmn1r103
- Entrez Gene ID
- 667129
- Gene Name
- vomeronasal 1 receptor 103
- Gene Aliases
- EG667129, Gm8474
- Chromosome Location
- Chr. 7: 20509617 - 20510534 on Build GRCm38
- UniGene ID
- Mm.480263
- Species
- Mus musculus
- Gene Symbol
- Vmn1r104
- Entrez Gene ID
- 667135
- Gene Name
- vomeronasal 1 receptor 104
- Gene Aliases
- EG667135, Gm8477
- Chromosome Location
- Chr. 7: 20533837 - 20534760 on Build GRCm38
- UniGene ID
- Mm.480262
- Species
- Mus musculus
- Gene Symbol
- Vmn1r107
- Entrez Gene ID
- 673977
- Gene Name
- vomeronasal 1 receptor 107
- Gene Aliases
- 100043561, Gm4519, Gm9609, Vmn1r110
- Chromosome Location
- Chr. 7: 20668918 - 20669841 on Build GRCm38
- UniGene ID
- Mm.480248
- Species
- Mus musculus
- Gene Symbol
- Vmn1r111
- Entrez Gene ID
- 667292
- Gene Name
- vomeronasal 1 receptor 111
- Gene Aliases
- EG667292, Gm8559
- Chromosome Location
- Chr. 7: 20702296 - 20703216 on Build GRCm38
- UniGene ID
- Mm.480254
- Species
- Mus musculus
- Gene Symbol
- Vmn1r112
- Entrez Gene ID
- 100043569
- Gene Name
- vomeronasal 1 receptor 112
- Gene Aliases
- EG667283, Gm16444
- Chromosome Location
- Chr. 7: 20771185 - 20772078 on Build GRCm38
- UniGene ID
- Mm.480299
- Species
- Mus musculus
- Gene Symbol
- Vmn1r114
- Entrez Gene ID
- 100042996
- Gene Name
- vomeronasal 1 receptor 114
- Gene Aliases
- ENSMUSG00000074323, Gm10669
- Chromosome Location
- Chr. 7: 20811221 - 20812186 on Build GRCm38
- UniGene ID
- Mm.480291
- Species
- Mus musculus
- Gene Symbol
- Vmn1r115
- Entrez Gene ID
- 667273
- Gene Name
- vomeronasal 1 receptor 115
- Gene Aliases
- EG667273, Gm8549
- Chromosome Location
- Chr. 7: 20844098 - 20844985 on Build GRCm38
- UniGene ID
- Mm.480255
- Species
- Mus musculus
- Gene Symbol
- Vmn1r117
- Entrez Gene ID
- 667262
- Gene Name
- vomeronasal 1 receptor 117
- Gene Aliases
- EG667262, Gm8544
- Chromosome Location
- Chr. 7: 20883198 - 20884121 on Build GRCm38
- UniGene ID
- Mm.480257
- Species
- Mus musculus
- Gene Symbol
- Vmn1r118
- Entrez Gene ID
- 667259
- Gene Name
- vomeronasal 1 receptor 118
- Gene Aliases
- EG667259, Gm8542
- Chromosome Location
- Chr. 7: 20911427 - 20912347 on Build GRCm38
- UniGene ID
- Mm.480258
- Species
- Mus musculus
- Gene Symbol
- Vmn1r119
- Entrez Gene ID
- 384696
- Gene Name
- vomeronasal 1 receptor 119
- Gene Aliases
- Gm1447
- Chromosome Location
- Chr. 7: 21011532 - 21012455 on Build GRCm38
- UniGene ID
- Mm.480283
- Species
- Mus musculus
- Gene Symbol
- Vmn1r121
- Entrez Gene ID
- 667240
- Gene Name
- vomeronasal 1 receptor 121
- Gene Aliases
- EG667240, Gm8533
- Chromosome Location
- Chr. 7: 21097569 - 21098513 on Build GRCm38
- UniGene ID
- Mm.480259
- Species
- Mus musculus
- Gene Symbol
- Vmn1r122
- Entrez Gene ID
- 435951
- Gene Name
- vomeronasal 1 receptor 122
- Gene Aliases
- EG435951, Gm5729
- Chromosome Location
- Chr. 7: 21133211 - 21134128 on Build GRCm38
- UniGene ID
- Mm.480280
- Species
- Mus musculus
- Gene Symbol
- Vmn1r124
- Entrez Gene ID
- 670764
- Gene Name
- vomeronasal 1 receptor 124
- Gene Aliases
- Gm9500
- Chromosome Location
- Chr. 7: 21259694 - 21260617 on Build GRCm38
- UniGene ID
- Mm.480250
- Species
- Mus musculus
- Gene Symbol
- Vmn1r125
- Entrez Gene ID
- 667215
- Gene Name
- vomeronasal 1 receptor 125
- Gene Aliases
- EG667215, Gm8519
- Chromosome Location
- Chr. 7: 21272179 - 21273102 on Build GRCm38
- UniGene ID
- Mm.480260
- Species
- Mus musculus
- Gene Symbol
- Vmn1r126
- Entrez Gene ID
- 100043013
- Gene Name
- vomeronasal 1 receptor 126
- Gene Aliases
- Gm4170
- Chromosome Location
- Chr. 7: 21300502 - 21301467 on Build GRCm38
- UniGene ID
- Mm.480289
- Species
- Mus musculus
- Gene Symbol
- Vmn1r127
- Entrez Gene ID
- 621561
- Gene Name
- vomeronasal 1 receptor 127
- Gene Aliases
- Gm6239
- Chromosome Location
- Chr. 7: 21318944 - 21319861 on Build GRCm38
- UniGene ID
- Mm.480270
- Species
- Mus musculus
- Gene Symbol
- Vmn1r128
- Entrez Gene ID
- 667199
- Gene Name
- vomeronasal 1 receptor 128
- Gene Aliases
- EG667199, Gm8509
- Chromosome Location
- Chr. 7: 21349373 - 21350296 on Build GRCm38
- UniGene ID
- Mm.480261
- Species
- Mus musculus
- Gene Symbol
- Vmn1r129
- Entrez Gene ID
- 621510
- Gene Name
- vomeronasal 1 receptor 129
- Gene Aliases
- EG621510, Gm6237
- Chromosome Location
- Chr. 7: 21360368 - 21361291 on Build GRCm38
- UniGene ID
- Mm.480271
- Species
- Mus musculus
- Gene Symbol
- Vmn1r130
- Entrez Gene ID
- 100043596
- Gene Name
- vomeronasal 1 receptor 130
- Gene Aliases
- Gm4542
- Chromosome Location
- Chr. 7: 21511780 - 21512703 on Build GRCm38
- UniGene ID
- Mm.484977
- Species
- Mus musculus
- Gene Symbol
- Vmn1r131
- Entrez Gene ID
- 100043016
- Gene Name
- vomeronasal 1 receptor 131
- Gene Aliases
- Gm4172
- Chromosome Location
- Chr. 7: 21545170 - 21546090 on Build GRCm38
- UniGene ID
- Mm.484991
- Species
- Mus musculus
- Gene Symbol
- Vmn1r132
- Entrez Gene ID
- 100043604
- Gene Name
- vomeronasal 1 receptor 132
- Gene Aliases
- EG667346, Gm16448
- Chromosome Location
- Chr. 7: 21612745 - 21615147 on Build GRCm38
- UniGene ID
- Mm.334360
- Species
- Mus musculus
- Gene Symbol
- Vmn1r135
- Entrez Gene ID
- 667404
- Gene Name
- vomeronasal 1 receptor 135
- Gene Aliases
- EG667404, Gm8611
- Chromosome Location
- Chr. 7: 21832351 - 21833274 on Build GRCm38
- UniGene ID
- Mm.484973
- Species
- Mus musculus
- Gene Symbol
- Vmn1r137
- Entrez Gene ID
- 100043614
- Gene Name
- vomeronasal 1 receptor 137
- Gene Aliases
- Gm4551
- Chromosome Location
- Chr. 7: 21994587 - 21995510 on Build GRCm38
- UniGene ID
- Mm.480298
- Species
- Mus musculus
- Gene Symbol
- Vmn1r138
- Entrez Gene ID
- 100043029
- Gene Name
- vomeronasal 1 receptor 138
- Gene Aliases
- Gm4179
- Chromosome Location
- Chr. 7: 22027977 - 22028897 on Build GRCm38
- UniGene ID
- Mm.482058
- Species
- Mus musculus
- Gene Symbol
- Vmn1r139
- Entrez Gene ID
- 667444
- Gene Name
- vomeronasal 1 receptor 139
- Gene Aliases
- EG667444, Gm8636
- Chromosome Location
- Chr. 7: 22096867 - 22097760 on Build GRCm38
- UniGene ID
- Mm.484972
- Species
- Mus musculus
- Gene Symbol
- Vmn1r142
- Entrez Gene ID
- 667464
- Gene Name
- vomeronasal 1 receptor 142
- Gene Aliases
- EG667464, Gm8647
- Chromosome Location
- Chr. 7: 22163118 - 22164035 on Build GRCm38
- UniGene ID
- Mm.480253
- Species
- Mus musculus
- Gene Symbol
- Vmn1r143
- Entrez Gene ID
- 667469
- Gene Name
- vomeronasal 1 receptor 143
- Gene Aliases
- EG667469, Gm8650
- Chromosome Location
- Chr. 7: 22189377 - 22190300 on Build GRCm38
- UniGene ID
- Mm.484971
- Species
- Mus musculus
- Gene Symbol
- Vmn1r148
- Entrez Gene ID
- 81011
- Gene Name
- vomeronasal 1 receptor 148
- Gene Aliases
- V1rd14, V3R8
- Chromosome Location
- Chr. 7: 22412231 - 22414669 on Build GRCm38
- UniGene ID
- Mm.160376
- Species
- Mus musculus
- Gene Symbol
- Vmn1r149
- Entrez Gene ID
- 100043051
- Gene Name
- vomeronasal 1 receptor 149
- Gene Aliases
- Gm4194
- Chromosome Location
- Chr. 7: 22437306 - 22438229 on Build GRCm38
- UniGene ID
- Mm.480287
- Species
- Mus musculus
- Gene Symbol
- Vmn1r151
- Entrez Gene ID
- 435947
- Gene Name
- vomeronasal 1 receptor 151
- Gene Aliases
- EG435947, Gm5727
- Chromosome Location
- Chr. 7: 22498761 - 22499678 on Build GRCm38
- UniGene ID
- Mm.480281
- Species
- Mus musculus
- Gene Symbol
- Vmn1r152
- Entrez Gene ID
- 667504
- Gene Name
- vomeronasal 1 receptor 152
- Gene Aliases
- EG667504, Gm8673
- Chromosome Location
- Chr. 7: 22522967 - 22523890 on Build GRCm38
- UniGene ID
- Mm.480252
- Species
- Mus musculus
- Gene Symbol
- Vmn1r155
- Entrez Gene ID
- 667530
- Gene Name
- vomeronasal 1 receptor 155
- Gene Aliases
- EG667530, Gm8687
- Chromosome Location
- Chr. 7: 22658018 - 22658941 on Build GRCm38
- UniGene ID
- Mm.484970
- Species
- Mus musculus
- Gene Symbol
- Vmn1r157
- Entrez Gene ID
- 667551
- Gene Name
- vomeronasal 1 receptor 157
- Gene Aliases
- 100043559, EG667551, Gm4517, Gm8699, Vmn1r109
- Chromosome Location
- Chr. 7: 22761697 - 22762590 on Build GRCm38
- UniGene ID
- Mm.484969
- Species
- Mus musculus
- Gene Symbol
- Vmn1r158
- Entrez Gene ID
- 100043067
- Gene Name
- vomeronasal 1 receptor 158
- Gene Aliases
- EG667554, Gm16455
- Chromosome Location
- Chr. 7: 22789859 - 22790782 on Build GRCm38
- UniGene ID
- Mm.480285
- Species
- Mus musculus
- Gene Symbol
- Vmn1r159
- Entrez Gene ID
- 670857
- Gene Name
- vomeronasal 1 receptor 159
- Gene Aliases
- ENSMUSG00000074298, Gm16507
- Chromosome Location
- Chr. 7: 22842688 - 22843605 on Build GRCm38
- UniGene ID
- Mm.480249
- Species
- Mus musculus
- Gene Symbol
- Vmn1r163
- Entrez Gene ID
- 667586
- Gene Name
- vomeronasal 1 receptor 163
- Gene Aliases
- EG667586, Gm8716
- Chromosome Location
- Chr. 7: 23055540 - 23056463 on Build GRCm38
- UniGene ID
- Mm.480251
- Species
- Mus musculus
- Gene Symbol
- Vmn1r165
- Entrez Gene ID
- 100043667
- Gene Name
- vomeronasal 1 receptor 165
- Gene Aliases
- Gm4579
- Chromosome Location
- Chr. 7: 23217332 - 23218255 on Build GRCm38
- UniGene ID
- Mm.480298
- Species
- Mus musculus
- Gene Symbol
- Vmn1r166
- Entrez Gene ID
- 100043088
- Gene Name
- vomeronasal 1 receptor 166
- Gene Aliases
- Gm4220
- Chromosome Location
- Chr. 7: 23250762 - 23251682 on Build GRCm38
- UniGene ID
- Mm.484991
- Species
- Mus musculus
- Gene Symbol
- Vmn1r178
- Entrez Gene ID
- 232959
- Gene Name
- vomeronasal 1 receptor 178
- Gene Aliases
- Gm473, V1rd13
- Chromosome Location
- Chr. 7: 23893529 - 23894443 on Build GRCm38
- UniGene ID
- Mm.377248
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC154405.1 |
- |
|
164 |
|
- Gene Symbol
- Vmn1r179
- Entrez Gene ID
- 404286
- Gene Name
- vomeronasal 1 receptor 179
- Gene Aliases
- V1rd17
- Chromosome Location
- Chr. 7: 23928386 - 23929357 on Build GRCm38
- UniGene ID
- Mm.377930
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_207545.1 |
NP_997428.1
|
1 |
209 |
|
- Gene Symbol
- Vmn1r180
- Entrez Gene ID
- 232962
- Gene Name
- vomeronasal 1 receptor 180
- Gene Aliases
- Gm475, V1rd16
- Chromosome Location
- Chr. 7: 23952387 - 23953356 on Build GRCm38
- UniGene ID
- Mm.377249
- Species
- Mus musculus
- Gene Symbol
- Vmn1r91
- Entrez Gene ID
- 667067
- Gene Name
- vomeronasal 1 receptor 91
- Gene Aliases
- EG667067, Gm8442
- Chromosome Location
- Chr. 7: 20101158 - 20102081 on Build GRCm38
- UniGene ID
- Mm.480264
- Species
- Mus musculus
- Gene Symbol
- Vmn1r93
- Entrez Gene ID
- 404290
- Gene Name
- vomeronasal 1 receptor 93
- Gene Aliases
- V1rd21
- Chromosome Location
- Chr. 7: 20138123 - 20139088 on Build GRCm38
- UniGene ID
- Mm.377932
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_207547.2 |
NP_997430.2
|
1 |
164 |
|
- Gene Symbol
- Vmn1r94
- Entrez Gene ID
- 620537
- Gene Name
- vomeronasal 1 receptor 94
- Gene Aliases
- EG620537, Gm6160
- Chromosome Location
- Chr. 7: 20167460 - 20168377 on Build GRCm38
- UniGene ID
- Mm.480273
- Species
- Mus musculus
- Gene Symbol
- Vmn1r95
- Entrez Gene ID
- 100312475
- Gene Name
- vomeronasal 1 receptor, 95
- Gene Aliases
- Gm17331
- Chromosome Location
- Chr. 7: 20193710 - 20194633 on Build GRCm38
- UniGene ID
- Mm.480294
- Species
- Mus musculus
View Full Product Details