Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133653.3 | NP_598414.1 | 10 | 3305 | |
GenBank mRNA | AK149542.1 | - | 9 | 3056 | |
AK153832.1 | - | 9 | 3059 | ||
BB807316.1 | - | 1 | 189 | ||
BC011211.1 | - | 9 | 3043 | 4196928 | |
L13622.1 | - | 9 | 3058 |