Gene Transcripts
- Gene Symbol
- Nalcn
- Entrez Gene ID
- 338370
- Gene Name
- sodium leak channel, non-selective
- Gene Aliases
- A530023G15Rik, AI849508, Vgcnl1
- Chromosome Location
- Chr. 14: 123276634 - 123627363 on Build GRCm38
- UniGene ID
- Mm.255348
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_177393.4 |
NP_796367.3
|
36, 37 |
4479 |
|
GenBank mRNA |
AK044431.1 |
- |
4, 5 |
426 |
|
View Full Product Details