Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001017916.1 | NP_001017916.1 | 5 | 720 | |
NM_001017917.1 | NP_001017917.1 | 5 | 553 | ||
NM_001915.3 | NP_001906.3 | 5 | 550 | ||
XM_005257091.1 | XP_005257148.1 | 5 | 495 | ||
XM_017024260.1 | XP_016879749.1 | 5 | 592 | ||
XM_017024261.1 | XP_016879750.1 | 4 | 433 | ||
XM_017024262.1 | XP_016879751.1 | 5 | 785 | ||
GenBank mRNA | AK294403.1 | - | 4 | 931 | |
AK294630.1 | - | 5 | 704 | ||
AK303177.1 | - | 5 | 419 | ||
AK316606.1 | - | 5 | 495 | ||
BC000021.1 | - | 5 | 581 | 3509626 | |
BC002976.1 | - | 5 | 507 | 3535771 | |
BC091485.1 | - | 5 | 521 | 6188259 | |
BF206825.1 | - | 556 | |||
BM552101.1 | - | 5 | 550 | ||
BT007096.1 | - | 4 | 420 |