Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007058.3 | NP_008989.2 | 7 | 815 | |
XM_006714984.1 | XP_006715047.1 | 7 | 818 | ||
XM_006714985.1 | XP_006715048.1 | 7 | 818 | ||
XM_006714987.1 | XP_006715050.1 | 7 | 818 | ||
XM_011514274.1 | XP_011512576.1 | 7 | 899 | ||
XM_011514275.1 | XP_011512577.1 | 8 | 941 | ||
GenBank mRNA | AJ242832.1 | - | 6 | 769 | |
AK314054.1 | - | 7 | 795 | ||
BC033733.1 | - | 7 | 786 | 5170266 |