Gene Transcripts
- Gene Symbol
- Enox2
- Entrez Gene ID
- 209224
- Gene Name
- ecto-NOX disulfide-thiol exchanger 2
- Gene Aliases
- APK1, Cova1, tNOX
- Chromosome Location
- Chr. X: 49009707 - 49288245 on Build GRCm38
- UniGene ID
- Mm.327730
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001271447.1 |
NP_001258376.1
|
14, 15 |
2011 |
|
NM_001271448.1 |
NP_001258377.1
|
13, 14 |
1856 |
|
NM_001271449.1 |
NP_001258378.1
|
14, 15 |
2000 |
|
NM_001271450.1 |
NP_001258379.1
|
14, 15 |
1967 |
|
NM_145951.5 |
NP_666063.1
|
14, 15 |
1955 |
|
GenBank mRNA |
AK034058.1 |
- |
14, 15 |
1956 |
|
AK041822.1 |
- |
14, 15 |
1968 |
|
AK133956.1 |
- |
13, 14 |
1827 |
|
AK137736.1 |
- |
14, 15 |
1982 |
|
AK139897.1 |
- |
14, 15 |
1982 |
|
BC025915.1 |
- |
14, 15 |
1974 |
5043650 |
View Full Product Details