Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001167976.1 | NP_001161448.1 | 7 | 454 | |
NM_172577.3 | NP_766165.2 | 6 | 897 | ||
GenBank mRNA | AK036988.1 | - | 6 | 637 | |
AK041694.1 | - | 6 | 898 | ||
AK045180.1 | - | 6 | 723 | ||
AK162379.1 | - | 6 | 724 | ||
AK167535.1 | - | 6 | 605 | ||
BC057980.1 | - | 6 | 721 | 5042653 | |
EU180236.1 | - | 7 | 454 |