Gene Transcripts
- Gene Symbol
- 4930590J08Rik
- Entrez Gene ID
- 381798
- Gene Name
- RIKEN cDNA 4930590J08 gene
- Gene Aliases
- Gm1071
- Chromosome Location
- Chr. 6: 91902692 - 91951736 on Build GRCm38
- UniGene ID
- Mm.444785
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_198668.2 |
NP_941070.1
|
20 |
3272 |
|
GenBank mRNA |
AK029822.1 |
- |
20 |
3272 |
|
View Full Product Details