Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159572.1 | NP_001153044.1 | 2 | 606 | |
NM_028732.4 | NP_083008.1 | 2 | 606 | ||
GenBank mRNA | AK014600.1 | - | 2 | 534 | |
AK030479.1 | - | 2 | 546 | ||
AK032383.1 | - | 2 | 551 | ||
AK033393.1 | - | 2 | 552 | ||
AK079734.1 | - | 2 | 554 | ||
AK131118.1 | - | 2 | 522 | ||
AK159086.1 | - | 2 | 551 | ||
AK159864.1 | - | 2 | 551 | ||
AK162635.1 | - | 445 | |||
AK163118.1 | - | 2 | 551 | ||
AK170059.1 | - | 2 | 554 | ||
AK170419.1 | - | 2 | 551 | ||
BC003967.1 | - | 2 | 522 | 3488482 | |
EF426799.1 | - | 2 | 435 | ||
JN602184.1 | - | 2 | 435 |