Gene Transcripts

Gene Symbol
Gm5136
Entrez Gene ID
368203
Gene Name
predicted gene 5136
Gene Aliases
C6.1AL, EG368203
Chromosome Location
Chr. 10: 108699044 - 108700160 on Build GRCm38
UniGene ID
Mm.452758
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203660.2 NP_988991.1 1 245
GenBank mRNA BC120506.1 - 1 245
BC120508.1 - 1 245 40129431

View Full Product Details