Gene Transcripts

Gene Symbol
Zbtb11
Entrez Gene ID
271377
Gene Name
zinc finger and BTB domain containing 11
Gene Aliases
9230110G02Rik, ZNF-U69274
Chromosome Location
Chr. 16: 55972697 - 56008913 on Build GRCm38
UniGene ID
Mm.282814
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173026.2 NP_766614.2 4 1483
GenBank mRNA AK033794.1 - 1 164
AK048360.1 - 1 1764

View Full Product Details