Gene Transcripts
- Gene Symbol
- Olfr1198
- Entrez Gene ID
- 404330
- Gene Name
- olfactory receptor 1198
- Gene Aliases
- GA_x5J8B7W62NC-67655-66851, GA_x6K02T2Q125-50221692-50220766, MOR225-13
- Chromosome Location
- Chr. 2: 88745960 - 88746886 on Build GRCm38
- UniGene ID
- Mm.377944
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_207567.1 |
NP_997450.1
|
1 |
187 |
|
View Full Product Details