Gene Transcripts

Gene Symbol
Adgrg3
Entrez Gene ID
54672
Gene Name
adhesion G protein-coupled receptor G3
Gene Aliases
A030001G24Rik, AW214383, Gpr97, Pb1, Pb99
Chromosome Location
Chr. 8: 95017114 - 95045250 on Build GRCm38
UniGene ID
Mm.27995
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173036.3 NP_766624.3 2 204
GenBank mRNA AK154648.1 - 2 180
AK155164.1 - 2 176
BC064726.1 - 2 164 30433392

View Full Product Details