Gene Transcripts

Gene Symbol
A930009A15Rik
Entrez Gene ID
77798
Gene Name
RIKEN cDNA A930009A15 gene
Gene Aliases
Chromosome Location
Chr. 10: 115569809 - 115582454 on Build GRCm38
UniGene ID
Mm.446685
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029982.1 NP_084258.1 1 247
GenBank mRNA AK020834.1 - 1 248

View Full Product Details