Gene Transcripts

Gene Symbol
Ints10
Entrez Gene ID
290679
Gene Name
integrator complex subunit 10
Gene Aliases
RGD1304711
Chromosome Location
Chr. 16: 22616480 - 22667762 on Build Rnor_6.0
UniGene ID
Rn.2385
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134416.1 NP_001127888.1 6, 7 700
XM_008771092.1 XP_008769314.1 6, 7 980
GenBank mRNA BC166856.1 - 6, 7 673

View Full Product Details