Gene Transcripts

Gene Symbol
Bcl2
Entrez Gene ID
24224
Gene Name
B-cell CLL/lymphoma 2
Gene Aliases
Bcl-2
Chromosome Location
Chr. 13: 26605426 - 26769374 on Build Rnor_6.0
UniGene ID
Rn.9996
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016993.1 NP_058689.1 1 750
GenBank mRNA AF149299.1 - 1 93
L14680.1 - 1 750
U34964.1 - 1 516

View Full Product Details