Gene Transcripts

Gene Symbol
Ctf1
Entrez Gene ID
29201
Gene Name
cardiotrophin 1
Gene Aliases
Chromosome Location
Chr. 1: 199162320 - 199168299 on Build Rnor_6.0
UniGene ID
Rn.10253
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017129.1 NP_058825.1 2 118
XM_006230221.2 XP_006230283.1 2 881
GenBank mRNA BC072690.1 - 2 107
D78591.1 - 2 118

View Full Product Details