Gene Transcripts

Gene Symbol
4930542D17Rik
Entrez Gene ID
75211
Gene Name
RIKEN cDNA 4930542D17 gene
Gene Aliases
-
Chromosome Location
Chr.16: 50590503 - 50654166 on Build GRCm38
Uni Gene
Mm.399337
Interrogated Sequence Translated Protein
RefSeq NR_040566.1
GenBank mRNA AK016020.1 -

View Full Product Details