Gene Transcripts
- Gene Symbol
- MAP3K14-AS1
- Entrez Gene ID
- 100133991
- Gene Name
- MAP3K14 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 17: 45247925 - 45268630 on Build GRCh38
- UniGene ID
- Hs.668927
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB103942.1 |
- |
3 |
549 |
|
Chr. 17 - 45247934 - 45268281 [+] |
GenBankmRNA |
DA712632.1 |
- |
3 |
549 |
|
Chr. 17 - 45247936 - 45268287 [+] |
View Full Product Details