Gene Transcripts
- Gene Symbol
- LOC102723448
- Entrez Gene ID
- 102723448
- Gene Name
- uncharacterized LOC102723448
- Gene Aliases
- Chromosome Location
- Chr. 3: 11751 - 24501 on Build GRCh38
- UniGene ID
- Hs.543290
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110824.1 |
|
4 |
195 |
|
GenBank mRNA |
BC039672.1 |
- |
4 |
195 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI826035.1 |
- |
4 |
194 |
|
Chr. 3 - 11749 - 24498 [+] |
View Full Product Details