Gene Transcripts
- Gene Symbol
- MCF2L
- Entrez Gene ID
- 23263
- Gene Name
- MCF.2 cell line derived transforming sequence like
- Gene Aliases
- ARHGEF14, DBS, OST
- Chromosome Location
- Chr. 13: 112894378 - 113099739 on Build GRCh38
- UniGene ID
- Hs.170422
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
W94194.1 |
- |
|
28 |
|
Chr. - - [-] |
GenBankmRNA |
BX111900.1 |
- |
1 |
29 |
|
Chr. 13 - 113099062 - 113099739 [+] |
View Full Product Details