Gene Transcripts
- Gene Symbol
- Gm35572
- Entrez Gene ID
- 102639212
- Gene Name
- predicted gene, 35572
- Gene Aliases
- Chromosome Location
- Chr. 8: 71753015 - 71756195 on Build GRCm38
- UniGene ID
- Mm.359424
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK142664.1 |
- |
|
1421 |
|
View Full Product Details