Gene Transcripts

Gene Symbol
Gm35572
Entrez Gene ID
102639212
Gene Name
predicted gene, 35572
Gene Aliases
Chromosome Location
Chr. 8: 71753015 - 71756195 on Build GRCm38
UniGene ID
Mm.359424
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK142664.1 - 547


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BY730903.1 - 4 547 Chr. 8 - 71753719 - 71781322 [-]

View Full Product Details